Assay Details
| Gene Symbol: DAAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006715039.3 | XP_006715102.3 | 16 - 17 | 2758 | 98 | |
| XM_006715042.2 | XP_006715105.1 | 16 - 17 | 4753 | 98 | ||
| XM_006715043.2 | XP_006715106.1 | 16 - 17 | 2130 | 98 | ||
| XM_006715045.3 | XP_006715108.1 | 16 - 17 | 3345 | 98 | ||
| XM_006715046.3 | XP_006715109.1 | 16 - 17 | 2523 | 98 | ||
| XM_017010630.1 | XP_016866119.1 | 16 - 17 | 3369 | 98 | ||
More Information
Set Membership: |
|

DAAM2
Genomic Map