Assay Details
Gene Symbol: ZW10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004724.3 | NP_004715.1 | 7 - 8 | 1017 | 67 | |
XM_017018559.1 | XP_016874048.1 | 7 - 8 | 973 | 67 | ||
GenBank mRNA | AK303871.1 | - | 6 - 7 | 696 | 67 | |
AK312925.1 | - | 7 - 8 | 995 | 67 | ||
BC128264.1 | - | 7 - 8 | 945 | 40118818 | 67 | |
U54996.1 | - | 7 - 8 | 943 | 67 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11002305.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic sister chromatid segregation
establishment of mitotic spindle orientation
protein complex assembly
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
Golgi organization
sister chromatid cohesion
mitotic metaphase plate congression
mitotic cell cycle checkpoint
mitotic spindle assembly checkpoint
regulation of exit from mitosis
meiotic nuclear division
protein transport
protein localization to kinetochore
cell division
protein binding
centromeric DNA binding
mitotic sister chromatid segregation
establishment of mitotic spindle orientation
protein complex assembly
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
Golgi organization
sister chromatid cohesion
mitotic metaphase plate congression
mitotic cell cycle checkpoint
mitotic spindle assembly checkpoint
regulation of exit from mitosis
meiotic nuclear division
protein transport
protein localization to kinetochore
cell division
protein binding
centromeric DNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027592.hg.1 | ENST00000200135.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11027592.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11027592.hg.1 | TC11002305.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100012353.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | ENST00000200135.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | ENST00000535142.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | ENST00000538209.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | NM_004724.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100194931.hg.1 | TC1100012353.hg.1 | Clariom D TC |
