Assay Details
| Gene Symbol: PSMD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005805.5 | NP_005796.1 | 6 - 7 | 783 | 89 | |
| GenBank mRNA | AK055128.1 | - | 6 - 7 | 1020 | 89 | |
| BC066336.1 | - | 6 - 7 | 629 | 5296432 | 89 | |
| U86782.1 | - | 6 - 7 | 515 | 89 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000970.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein polyubiquitination
double-strand break repair via homologous recombination
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
double-strand break repair via nonhomologous end joining
ubiquitin-dependent protein catabolic process
regulation of cellular amino acid metabolic process
positive regulation of endopeptidase activity
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
response to ethanol
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
regulation of proteasomal protein catabolic process
protein K63-linked deubiquitination
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
protein binding
metallopeptidase activity
metal ion binding
endopeptidase activator activity
proteasome binding
MAPK cascade
protein polyubiquitination
double-strand break repair via homologous recombination
stimulatory C-type lectin receptor signaling pathway
antigen processing and presentation of exogenous peptide antigen via MHC class I, TAP-dependent
double-strand break repair via nonhomologous end joining
ubiquitin-dependent protein catabolic process
regulation of cellular amino acid metabolic process
positive regulation of endopeptidase activity
anaphase-promoting complex-dependent catabolic process
tumor necrosis factor-mediated signaling pathway
NIK/NF-kappaB signaling
Fc-epsilon receptor signaling pathway
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of mRNA stability
response to ethanol
T cell receptor signaling pathway
negative regulation of ubiquitin-protein ligase activity involved in mitotic cell cycle
positive regulation of ubiquitin-protein ligase activity involved in regulation of mitotic cell cycle transition
Wnt signaling pathway, planar cell polarity pathway
regulation of proteasomal protein catabolic process
protein K63-linked deubiquitination
negative regulation of canonical Wnt signaling pathway
positive regulation of canonical Wnt signaling pathway
protein binding
metallopeptidase activity
metal ion binding
endopeptidase activator activity
proteasome binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02014234.hg.1 | TC02000970.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014234.hg.1 | ENST00000409682.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02014234.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000478123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000478123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000478123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000478123.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000492908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000492908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000492908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | ENST00000492908.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200168495.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200168495.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200168495.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200168495.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200168495.hg.1 | TC0200009813.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000409682.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000437630.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000477232.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000478123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000478123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000478123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000478123.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000492908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000492908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000492908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | ENST00000492908.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009813.hg.1 | NM_005805.5 | Refseq |

PSMD14
Genomic Map