Assay Details
| Gene Symbol: PSMD14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005805.5 | NP_005796.1 | 9 - 10 | 1112 | 132 | |
| GenBank mRNA | AB209220.1 | - | 2 - 3 | 4288 | 132 | |
| AK055128.1 | - | 9 - 10 | 1349 | 132 | ||
| BC066336.1 | - | 9 - 10 | 958 | 5296432 | 132 | |
| BM978668.1 | - | 3 - 4 | 132 | |||
| U86782.1 | - | 9 - 10 | 844 | 132 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000970.hg.1
Set Membership: |
|

PSMD14
Genomic Map