Assay Details
| Gene Symbol: UNC5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133369.2 | NP_588610.2 | 1 - 2 | 265 | 123 | |
| XM_006714927.1 | XP_006714990.1 | 1 - 2 | 245 | 123 | ||
| XM_006714928.1 | XP_006714991.1 | 1 - 2 | 245 | 123 | ||
| XM_011534686.1 | XP_011532988.1 | 1 - 2 | 245 | 123 | ||
| XM_011534687.2 | XP_011532989.1 | 1 - 2 | 245 | 123 | ||
| GenBank mRNA | AK131380.1 | - | 2 - 3 | 142 | 123 | |
| BC157824.1 | - | 1 - 2 | 148 | 9057043 | 123 | |
| BI818609.1 | - | 1 - 2 | 76 | 123 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001007.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05014506.hg.1 | TC05001007.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014506.hg.1 | ENST00000329542.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014506.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014505.hg.1 | TC05001007.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014505.hg.1 | ENST00000329542.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014505.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014508.hg.1 | TC05001007.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014508.hg.1 | ENST00000329542.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014508.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014509.hg.1 | TC05001007.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014509.hg.1 | ENST00000329542.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014509.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014507.hg.1 | TC05001007.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014507.hg.1 | ENST00000329542.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05014507.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167286.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167286.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167286.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167286.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167286.hg.1 | TC0500009603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009603.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167284.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167284.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167284.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167284.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167284.hg.1 | TC0500009603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | ENST00000509580.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167288.hg.1 | TC0500009603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500167287.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167287.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167287.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167287.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167287.hg.1 | ENST00000513890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167287.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167287.hg.1 | TC0500009603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500167283.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167283.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167283.hg.1 | ENST00000329542.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500167283.hg.1 | NM_133369.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500167283.hg.1 | TC0500009603.hg.1 | Clariom D TC |

UNC5A
Genomic Map