| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
TC04000469.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
ENST00000316707.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
ENST00000357349.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
ENST00000411767.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
ENST00000427191.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
ENST00000436978.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
ENST00000511467.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
NM_006264.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
NM_080683.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
NM_080684.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04006737.hg.1 |
NM_080685.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000316707.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000411767.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000427191.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000436978.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000511467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
NM_006264.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
NM_080683.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
NM_080684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
NM_080685.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000316707.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000411767.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000427191.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000436978.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
ENST00000511467.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
NM_006264.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
NM_080683.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
NM_080684.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
NM_080685.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400155338.hg.1 |
TC0400008041.hg.1 |
Clariom D TC |