Assay Details
| Gene Symbol: CEBPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005760.2 | NP_005751.2 | 3 - 4 | 1912 | 157 | |
| GenBank mRNA | AK312322.1 | - | 3 - 4 | 1911 | 157 | |
| BC034475.2 | - | 3 - 4 | 1916 | 4822708 | 157 | |
| M37197.1 | - | 3 - 4 | 1893 | 157 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001741.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
poly(A) RNA binding
transcription from RNA polymerase II promoter
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02028051.hg.1 | TC02001741.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02028051.hg.1 | ENST00000234170.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02028051.hg.1 | ENST00000545744.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02028051.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000446769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000446769.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189498.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189498.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189498.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189498.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189498.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200189498.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200189498.hg.1 | TC0200012260.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000234170.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000446769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000446769.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000489306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | ENST00000619061.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012260.hg.1 | NM_005760.2 | Refseq |

CEBPZ
Genomic Map