Assay Details
| Gene Symbol: DBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016216.3 | NP_057300.2 | 2 - 3 | 475 | 80 | |
| GenBank mRNA | AF180919.2 | - | 2 - 3 | 429 | 80 | |
| AK094147.1 | - | 3 - 4 | 360 | 80 | ||
| AL702928.1 | - | 2 - 3 | 477 | 80 | ||
| BC009472.1 | - | 2 - 3 | 403 | 3503627 | 80 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001821.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK094147.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03032945.hg.1 | TC03001821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032945.hg.1 | ENST00000260803.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032945.hg.1 | ENST00000505015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032945.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032951.hg.1 | TC03001821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032951.hg.1 | ENST00000260803.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032951.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032951.hg.1 | ENST00000505015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032951.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032952.hg.1 | TC03001821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032952.hg.1 | ENST00000260803.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032952.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032952.hg.1 | ENST00000505015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032952.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032947.hg.1 | TC03001821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032947.hg.1 | ENST00000260803.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032947.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032947.hg.1 | ENST00000505015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032947.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032949.hg.1 | TC03001821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032949.hg.1 | ENST00000260803.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032949.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032949.hg.1 | ENST00000505015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032949.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032948.hg.1 | TC03001821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032948.hg.1 | ENST00000260803.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032948.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032948.hg.1 | ENST00000505015.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032948.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032950.hg.1 | TC03001821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032950.hg.1 | ENST00000260803.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032950.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032950.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032955.hg.1 | TC03001821.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032955.hg.1 | ENST00000260803.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03032955.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000460271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000460271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000460271.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193942.hg.1 | TC0300012567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193941.hg.1 | TC0300012567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193943.hg.1 | TC0300012567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193946.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193946.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193946.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193946.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193946.hg.1 | TC0300012567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193938.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193938.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193938.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193938.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193938.hg.1 | TC0300012567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300193940.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193940.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193940.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193940.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193940.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300193940.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300193940.hg.1 | TC0300012567.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000260803.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000460271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000460271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000460271.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000463982.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | ENST00000477557.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012567.hg.1 | NM_016216.3 | Refseq |

DBR1
Genomic Map