Assay Details
| Gene Symbol: CTNNBIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020248.2 | NP_064633.1 | 2 - 3 | 195 | 63 | |
| XM_017001855.1 | XP_016857344.1 | 1 - 2 | 137 | 63 | ||
| GenBank mRNA | AB021262.1 | - | 2 - 3 | 161 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002187.hg.1
Additional Information:
This assay is in the 5'UTR region of the AB021262.1,NM_020248.2,XM_017001855.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
branching involved in ureteric bud morphogenesis
regulation of vascular permeability involved in acute inflammatory response
anterior/posterior pattern specification
Wnt signaling pathway
negative regulation of Wnt signaling pathway
negative regulation of protein binding
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of monocyte differentiation
positive regulation of osteoblast differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of transcription initiation from RNA polymerase II promoter
negative regulation of mesenchymal cell proliferation
protein binding
beta-catenin binding
armadillo repeat domain binding
branching involved in ureteric bud morphogenesis
regulation of vascular permeability involved in acute inflammatory response
anterior/posterior pattern specification
Wnt signaling pathway
negative regulation of Wnt signaling pathway
negative regulation of protein binding
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of monocyte differentiation
positive regulation of osteoblast differentiation
negative regulation of smooth muscle cell proliferation
negative regulation of transcription initiation from RNA polymerase II promoter
negative regulation of mesenchymal cell proliferation
protein binding
beta-catenin binding
armadillo repeat domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01033683.hg.1 | TC01002187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033683.hg.1 | ENST00000377263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033683.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033685.hg.1 | TC01002187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033685.hg.1 | ENST00000377263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033685.hg.1 | ENST00000400904.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033685.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033685.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033681.hg.1 | TC01002187.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033681.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033681.hg.1 | ENST00000377258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033681.hg.1 | ENST00000377263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033681.hg.1 | ENST00000400904.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033681.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01033681.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194747.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194747.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194747.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194747.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194747.hg.1 | TC0100012818.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194749.hg.1 | TC0100012818.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100012818.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | ENST00000377256.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | ENST00000377258.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | ENST00000377263.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | ENST00000400904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | NM_001012329.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | NM_020248.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100194745.hg.1 | TC0100012818.hg.1 | Clariom D TC |

CTNNBIP1
Genomic Map