Assay Details
| Gene Symbol: CAMK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003656.4 | NP_003647.1 | 4 - 5 | 475 | 84 | |
| XM_005265516.1 | XP_005265573.1 | 4 - 5 | 429 | 84 | ||
| XM_005265517.2 | XP_005265574.1 | 3 - 4 | 346 | 84 | ||
| XM_017007354.1 | XP_016862843.1 | 3 - 4 | 346 | 84 | ||
| GenBank mRNA | AB451277.1 | - | 3 - 4 | 290 | 84 | |
| AB451407.1 | - | 3 - 4 | 290 | 84 | ||
| AK314917.1 | - | 4 - 5 | 429 | 84 | ||
| BC106754.2 | - | 4 - 5 | 436 | 40024362 | 84 | |
| BC106755.2 | - | 4 - 5 | 436 | 40024363 | 84 | |
| BI546222.1 | - | 4 - 5 | 473 | 84 | ||
| DN990042.1 | - | 4 - 5 | 488 | 84 | ||
| L41816.1 | - | 4 - 5 | 437 | 84 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001149.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
nucleocytoplasmic transport
cell cycle
signal transduction
positive regulation of neuron projection development
negative regulation of protein binding
regulation of protein localization
positive regulation of peptidyl-serine phosphorylation
regulation of protein binding
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
regulation of muscle cell differentiation
positive regulation of muscle cell differentiation
positive regulation of synapse structural plasticity
positive regulation of syncytium formation by plasma membrane fusion
positive regulation of dendritic spine development
positive regulation of protein serine/threonine kinase activity
positive regulation of protein acetylation
regulation of histone H3-K9 acetylation
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
protein phosphorylation
nucleocytoplasmic transport
cell cycle
signal transduction
positive regulation of neuron projection development
negative regulation of protein binding
regulation of protein localization
positive regulation of peptidyl-serine phosphorylation
regulation of protein binding
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
regulation of muscle cell differentiation
positive regulation of muscle cell differentiation
positive regulation of synapse structural plasticity
positive regulation of syncytium formation by plasma membrane fusion
positive regulation of dendritic spine development
positive regulation of protein serine/threonine kinase activity
positive regulation of protein acetylation
regulation of histone H3-K9 acetylation
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03019822.hg.1 | TC03001149.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019822.hg.1 | ENST00000256460.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03019822.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300174407.hg.1 | TC0300010208.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000256460.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000397277.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000411972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000421120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000482803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | ENST00000496534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300010208.hg.1 | NM_003656.4 | Refseq |

CAMK1
Genomic Map