Assay Details
| Gene Symbol: TNFRSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243.4 | NP_001234.3 | 14 - 15 | 1764 | 87 | |
| NM_001281430.2 | NP_001268359.2 | 13 - 14 | 1673 | 87 | ||
| XM_011542441.2 | XP_011540743.1 | 14 - 15 | 1820 | 87 | ||
| XM_011542443.2 | XP_011540745.1 | 13 - 14 | 1841 | 87 | ||
| GenBank mRNA | BC063482.1 | - | 4 - 5 | 439 | 87 | |
| BC136400.1 | - | 13 - 14 | 1580 | 9020387 | 87 | |
| BC141804.1 | - | 5 - 6 | 319 | 87 | ||
| BC171762.1 | - | 13 - 14 | 1580 | 87 | ||
| D86042.1 | - | 5 - 6 | 441 | 87 | ||
| M83554.1 | - | 14 - 15 | 1764 | 87 | ||
| S75768.1 | - | 14 - 15 | 1542 | 87 | ||
More Information
Set Membership: |
|

TNFRSF8
Genomic Map