Assay Details
| Gene Symbol: LAX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136190.1 | NP_001129662.1 | 3 - 4 | 450 | 89 | |
| NM_001282878.1 | NP_001269807.1 | 3 - 4 | 281 | 89 | ||
| NM_017773.3 | NP_060243.2 | 3 - 4 | 721 | 89 | ||
| XM_006711397.3 | XP_006711460.1 | 4 - 5 | 612 | 89 | ||
| GenBank mRNA | AK000347.1 | - | 3 - 4 | 283 | 89 | |
| AK225331.1 | - | 3 - 4 | 283 | 89 | ||
| AK301076.1 | - | 3 - 4 | 701 | 89 | ||
| AK301421.1 | - | 3 - 4 | 423 | 89 | ||
| AY090784.1 | - | 3 - 4 | 631 | 89 | ||
| BC069650.1 | - | 2 - 3 | 167 | 7262467 | 89 | |
| BC089408.1 | - | 3 - 4 | 668 | 3092611 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001691.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
adaptive immune response
immune response
intracellular signal transduction
B cell activation
immunoglobulin secretion
antigen receptor-mediated signaling pathway
negative regulation of T cell activation
protein binding
protein kinase binding
SH2 domain binding
inactivation of MAPK activity
adaptive immune response
immune response
intracellular signal transduction
B cell activation
immunoglobulin secretion
antigen receptor-mediated signaling pathway
negative regulation of T cell activation
protein binding
protein kinase binding
SH2 domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01026265.hg.1 | TC01001691.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026265.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026265.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026265.hg.1 | ENST00000442561.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026265.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01026265.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100182781.hg.1 | TC0100011270.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000367217.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | ENST00000442561.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001136190.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_001282878.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011270.hg.1 | NM_017773.3 | Refseq |

LAX1
Genomic Map