| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022489.hg.1 |
TC09001748.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022489.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022489.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR09022489.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182562.hg.1 |
TC0900012024.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0900012024.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
ENST00000473911.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
ENST00000480181.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
ENST00000494426.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
NM_004669.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0900182559.hg.1 |
TC0900012024.hg.1 |
Clariom D TC |