Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253830.1 | NP_001240759.1 | 1 - 2 | 323 | 72 | |
| NM_177995.2 | NP_818931.1 | 1 - 2 | 338 | 72 | ||
| XM_017014282.1 | XP_016869771.1 | 1 - 2 | 541 | 72 | ||
| GenBank mRNA | AY171233.1 | - | 1 - 2 | 343 | 72 | |
| BC067120.1 | - | 1 - 2 | 323 | 5539182 | 72 | |
| BI460503.1 | - | 1 - 2 | 343 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000455.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY171233.1,BC067120.1,NM_001253830.1,NM_177995.2,XM_017014282.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09004893.hg.1 | TC09000455.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004893.hg.1 | ENST00000375360.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004893.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004893.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004892.hg.1 | TC09000455.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004892.hg.1 | ENST00000375360.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004892.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09004892.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153934.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153934.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153934.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153934.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153934.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153934.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153934.hg.1 | TC0900008010.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900153932.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153932.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153932.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153932.hg.1 | ENST00000375360.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900153932.hg.1 | NM_001253830.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153932.hg.1 | NM_177995.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900153932.hg.1 | TC0900008010.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 128960 | - | - |

PTPDC1
Genomic Map