Assay Details
| Gene Symbol: DUSP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004090.3 | NP_004081.1 | 1 - 2 | 197 | 73 | |
| GenBank mRNA | AB451424.1 | - | 1 - 2 | 121 | 73 | |
| AK129822.1 | - | 1 - 2 | 142 | 73 | ||
| BC002682.1 | - | 1 - 2 | 176 | 3605391 | 73 | |
| BT019522.1 | - | 1 - 2 | 121 | 73 | ||
| BU177084.1 | - | 1 - 2 | 177 | 73 | ||
| DA104250.1 | - | 1 - 2 | 197 | 73 | ||
| L05147.1 | - | 1 - 2 | 149 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001563.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
inactivation of MAPK activity
dephosphorylation
peptidyl-tyrosine dephosphorylation
negative regulation of MAPK cascade
positive regulation of mitotic cell cycle
negative regulation of JNK cascade
negative regulation of T cell receptor signaling pathway
negative regulation of T cell activation
negative regulation of ERK1 and ERK2 cascade
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity
protein kinase binding
MAP kinase phosphatase activity
inactivation of MAPK activity
dephosphorylation
peptidyl-tyrosine dephosphorylation
negative regulation of MAPK cascade
positive regulation of mitotic cell cycle
negative regulation of JNK cascade
negative regulation of T cell receptor signaling pathway
negative regulation of T cell activation
negative regulation of ERK1 and ERK2 cascade
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
phosphatase activity
protein kinase binding
MAP kinase phosphatase activity
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17020937.hg.1 | ENST00000226004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020937.hg.1 | ENST00000397937.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020937.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020937.hg.1 | TC17001563.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020942.hg.1 | ENST00000226004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020942.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020942.hg.1 | TC17001563.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020941.hg.1 | ENST00000226004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020941.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020941.hg.1 | TC17001563.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020939.hg.1 | ENST00000226004.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020939.hg.1 | ENST00000397937.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020939.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020939.hg.1 | TC17001563.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000591618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | ENST00000591618.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008785.hg.1 | ENST00000438334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590753.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000591618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | ENST00000591618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010798.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186102.hg.1 | TC1700010798.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700186110.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186110.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186110.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186110.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186110.hg.1 | TC1700010798.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700186108.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186108.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186108.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186108.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186108.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186108.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186108.hg.1 | TC1700010798.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500008785.hg.1 | ENST00000438334.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | ENST00000591618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | ENST00000226004.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | ENST00000590342.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | ENST00000590935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | ENST00000591618.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | NM_004090.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700186103.hg.1 | TC1700010798.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 161352 | - | - |

DUSP3
Genomic Map