Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000475574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000475574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000475574.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004702.hg.1 |
TC09000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004702.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004713.hg.1 |
TC09000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004713.hg.1 |
ENST00000375464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004713.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004714.hg.1 |
TC09000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004714.hg.1 |
ENST00000375464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004714.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004712.hg.1 |
TC09000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004712.hg.1 |
ENST00000375464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004712.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004706.hg.1 |
TC09000442.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004706.hg.1 |
ENST00000375464.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09004706.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153687.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153687.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153687.hg.1 |
TC0900007977.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900153699.hg.1 |
TC0900007977.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000375464.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466409.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000466929.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000468781.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000475574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000475574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000475574.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000488630.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000490488.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000495641.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
ENST00000498243.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900007977.hg.1 |
NM_032310.4 |
Refseq |