Assay Details
| Gene Symbol: HELZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014877.3 | NP_055692.2 | 2 - 3 | 103 | 125 | |
| XM_005257888.4 | XP_005257945.1 | 2 - 3 | 151 | 125 | ||
| XM_005257889.4 | XP_005257946.1 | 2 - 3 | 151 | 125 | ||
| XM_006722214.3 | XP_006722277.1 | 2 - 3 | 151 | 125 | ||
| XM_006722215.3 | XP_006722278.1 | 2 - 3 | 151 | 125 | ||
| XM_017025477.1 | XP_016880966.1 | 2 - 3 | 151 | 125 | ||
| XM_017025478.1 | XP_016880967.1 | 2 - 3 | 151 | 125 | ||
| GenBank mRNA | BC130582.1 | - | 2 - 3 | 83 | 40146613 | 125 |
| BC144083.1 | - | 2 - 3 | 83 | 9052603 | 125 | |
| BP353830.1 | - | 2 - 3 | 103 | 125 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC130582.1, BC144083.1, NM_014877.3, XM_005257888.4, XM_005257889.4, XM_006722214.3, XM_006722215.3, XM_017025477.1, XM_017025478.1 transcripts.
Set Membership: |
|

HELZ
Genomic Map