Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002266.3 | NP_002257.1 | 1 - 2 | 238 | 69 | |
| GenBank mRNA | AK024833.1 | - | 1 - 2 | 106 | 69 | |
| AK058111.1 | - | 1 - 2 | 107 | 69 | ||
| AK291954.1 | - | 1 - 2 | 105 | 69 | ||
| BC005978.1 | - | 1 - 2 | 105 | 4105058 | 69 | |
| BC013257.1 | - | 1 - 2 | 101 | 69 | ||
| BC034443.1 | - | 1 - 2 | 64 | 69 | ||
| BC053343.1 | - | 1 - 2 | 94 | 4450470 | 69 | |
| BC146905.1 | - | 1 - 2 | 92 | 9056833 | 69 | |
| DC388846.1 | - | 1 - 2 | 69 | |||
| U09559.1 | - | 1 - 2 | 108 | 69 | ||
| U28386.1 | - | 1 - 2 | 108 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK291954.1, BC005978.1, BC053343.1, BC146905.1, NM_002266.3, U09559.1, U28386.1 transcripts.
Set Membership: |
|

KPNA2
Genomic Map