Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320611.1 | NP_001307540.1 | 3 - 4 | 830 | 88 | |
| NM_002266.3 | NP_002257.1 | 3 - 4 | 472 | 88 | ||
| GenBank mRNA | AK024833.1 | - | 3 - 4 | 340 | 88 | |
| AK291954.1 | - | 3 - 4 | 339 | 88 | ||
| BC005978.1 | - | 3 - 4 | 339 | 4105058 | 88 | |
| BC013257.1 | - | 3 - 4 | 335 | 88 | ||
| BC034443.1 | - | 3 - 4 | 298 | 88 | ||
| BC053343.1 | - | 3 - 4 | 328 | 4450470 | 88 | |
| BC067848.1 | - | 3 - 4 | 571 | 4825235 | 88 | |
| BC146905.1 | - | 3 - 4 | 326 | 9056833 | 88 | |
| BT006665.1 | - | 2 - 3 | 210 | 88 | ||
| DC388846.1 | - | 3 - 4 | 472 | 88 | ||
| U09559.1 | - | 3 - 4 | 342 | 88 | ||
| U28386.1 | - | 3 - 4 | 342 | 88 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map