Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320611.1 | NP_001307540.1 | 5 - 6 | 1191 | 79 | |
| NM_002266.3 | NP_002257.1 | 5 - 6 | 833 | 79 | ||
| GenBank mRNA | AK024833.1 | - | 5 - 6 | 701 | 79 | |
| AK058111.1 | - | 5 - 6 | 702 | 79 | ||
| AK291954.1 | - | 5 - 6 | 700 | 79 | ||
| BC005978.1 | - | 5 - 6 | 700 | 4105058 | 79 | |
| BC013257.1 | - | 5 - 6 | 696 | 79 | ||
| BC034443.1 | - | 5 - 6 | 659 | 79 | ||
| BC053343.1 | - | 5 - 6 | 689 | 4450470 | 79 | |
| BC146905.1 | - | 5 - 6 | 687 | 9056833 | 79 | |
| BT006665.1 | - | 4 - 5 | 571 | 79 | ||
| U09559.1 | - | 5 - 6 | 703 | 79 | ||
| U28386.1 | - | 5 - 6 | 703 | 79 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map