Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320611.1 | NP_001307540.1 | 7 - 8 | 1550 | 97 | |
| NM_002266.3 | NP_002257.1 | 7 - 8 | 1192 | 97 | ||
| GenBank mRNA | AK024833.1 | - | 7 - 8 | 1060 | 97 | |
| AK058111.1 | - | 7 - 8 | 1061 | 97 | ||
| AK291954.1 | - | 7 - 8 | 1059 | 97 | ||
| BC005978.1 | - | 7 - 8 | 1059 | 4105058 | 97 | |
| BC013257.1 | - | 7 - 8 | 1055 | 97 | ||
| BC034443.1 | - | 7 - 8 | 1018 | 97 | ||
| BC053343.1 | - | 7 - 8 | 1048 | 4450470 | 97 | |
| BC067848.1 | - | 7 - 8 | 1291 | 4825235 | 97 | |
| BC146905.1 | - | 7 - 8 | 1046 | 9056833 | 97 | |
| BT006665.1 | - | 6 - 7 | 930 | 97 | ||
| U09559.1 | - | 7 - 8 | 1062 | 97 | ||
| U28386.1 | - | 7 - 8 | 1062 | 97 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map