Assay Details
| Gene Symbol: KPNA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320611.1 | NP_001307540.1 | 8 - 9 | 1786 | 97 | |
| NM_002266.3 | NP_002257.1 | 8 - 9 | 1428 | 97 | ||
| GenBank mRNA | AK024833.1 | - | 8 - 9 | 1296 | 97 | |
| AK058111.1 | - | 8 - 9 | 1297 | 97 | ||
| AK291954.1 | - | 8 - 9 | 1295 | 97 | ||
| BC005978.1 | - | 8 - 9 | 1295 | 4105058 | 97 | |
| BC013257.1 | - | 8 - 9 | 1291 | 97 | ||
| BC034443.1 | - | 8 - 9 | 1254 | 97 | ||
| BC053343.1 | - | 8 - 9 | 1284 | 4450470 | 97 | |
| BC067848.1 | - | 8 - 9 | 1527 | 4825235 | 97 | |
| BC146905.1 | - | 8 - 9 | 1282 | 9056833 | 97 | |
| BT006665.1 | - | 7 - 8 | 1166 | 97 | ||
| U09559.1 | - | 8 - 9 | 1298 | 97 | ||
| U28386.1 | - | 8 - 9 | 1298 | 97 | ||
More Information
Set Membership: |
|

KPNA2
Genomic Map