Assay Details
| Gene Symbol: CENPI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011530897.2 | XP_011529199.1 | 3 - 4 | 315 | 81 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011530897.2 transcripts.
Set Membership: |
|

CENPI
Genomic Map