Assay Details
| Gene Symbol: MZT2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005263742.2 | XP_005263799.1 | 2 - 3 | 658 | 76 | |
| Gene Symbol: MZT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011511889.2 | XP_011510191.1 | 2 - 3 | 414 | 76 | |
More Information
Set Membership: |
|

MZT2A
Genomic Map