| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000343242.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000403150.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
ENST00000406174.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700006520.hg.1 |
NM_002360.3 |
Refseq |