Assay Details
| Gene Symbol: OGG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016826.2 | NP_058434.1 | 4 - 5 | 1091 | 69 | |
| GenBank mRNA | AB019528.1 | - | 3 - 4 | 464 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: OGG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016826.2 | NP_058434.1 | 4 - 5 | 1091 | 69 | |
| GenBank mRNA | AB019528.1 | - | 3 - 4 | 464 | 69 | |
Set Membership: |
|