Assay Details
| Gene Symbol: DLL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019074.3 | NP_061947.1 | 1 - 2 | 402 | 83 | |
| GenBank mRNA | AB036931.1 | - | 1 - 2 | 342 | 83 | |
| AB043894.1 | - | 1 - 2 | 66 | 83 | ||
| AF253468.1 | - | 1 - 2 | 66 | 83 | ||
| AK308755.1 | - | 1 - 2 | 395 | 83 | ||
| AK313831.1 | - | 1 - 2 | 397 | 83 | ||
| AY358894.1 | - | 1 - 2 | 101 | 83 | ||
| BC106950.2 | - | 1 - 2 | 202 | 40034887 | 83 | |
| DA089757.1 | - | 1 - 2 | 402 | 83 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000299.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
patterning of blood vessels
blood vessel remodeling
cardiac ventricle morphogenesis
cardiac atrium morphogenesis
ventricular trabecula myocardium morphogenesis
pericardium morphogenesis
signal transduction
Notch signaling pathway
Notch receptor processing
visual perception
blood circulation
negative regulation of cell proliferation
negative regulation of endothelial cell migration
positive regulation of gene expression
negative regulation of gene expression
T cell differentiation
dorsal aorta morphogenesis
cellular response to vascular endothelial growth factor stimulus
cellular response to fibroblast growth factor stimulus
negative regulation of Notch signaling pathway
positive regulation of Notch signaling pathway
regulation of neurogenesis
ventral spinal cord interneuron fate commitment
regulation of neural retina development
Notch signaling involved in heart development
blood vessel lumenization
negative regulation of cell migration involved in sprouting angiogenesis
negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
positive regulation of neural precursor cell proliferation
Notch binding
calcium ion binding
protein binding
negative regulation of transcription from RNA polymerase II promoter
angiogenesis
patterning of blood vessels
blood vessel remodeling
cardiac ventricle morphogenesis
cardiac atrium morphogenesis
ventricular trabecula myocardium morphogenesis
pericardium morphogenesis
signal transduction
Notch signaling pathway
Notch receptor processing
visual perception
blood circulation
negative regulation of cell proliferation
negative regulation of endothelial cell migration
positive regulation of gene expression
negative regulation of gene expression
T cell differentiation
dorsal aorta morphogenesis
cellular response to vascular endothelial growth factor stimulus
cellular response to fibroblast growth factor stimulus
negative regulation of Notch signaling pathway
positive regulation of Notch signaling pathway
regulation of neurogenesis
ventral spinal cord interneuron fate commitment
regulation of neural retina development
Notch signaling involved in heart development
blood vessel lumenization
negative regulation of cell migration involved in sprouting angiogenesis
negative regulation of blood vessel endothelial cell proliferation involved in sprouting angiogenesis
positive regulation of neural precursor cell proliferation
Notch binding
calcium ion binding
protein binding
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15002127.hg.1 | ENST00000249749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002127.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002127.hg.1 | TC15000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002124.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002124.hg.1 | TC15000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002125.hg.1 | ENST00000249749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002125.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002125.hg.1 | TC15000299.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002126.hg.1 | ENST00000249749.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002126.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15002126.hg.1 | TC15000299.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500148774.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148774.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148774.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148774.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148774.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148774.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148774.hg.1 | TC1500006981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500148776.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148776.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148776.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148776.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148776.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148776.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148776.hg.1 | TC1500006981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500148777.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148777.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148777.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148777.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148777.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148777.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148777.hg.1 | TC1500006981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500148773.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148773.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148773.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148773.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148773.hg.1 | TC1500006981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500148775.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148775.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148775.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148775.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148775.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500148775.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500148775.hg.1 | TC1500006981.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000249749.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000557876.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | ENST00000559440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500006981.hg.1 | NM_019074.3 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 159761 | - | - |

DLL4
Genomic Map