Assay Details
| Gene Symbol: GNB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021629.3 | NP_067642.1 | 8 - 9 | 974 | 62 | |
| XM_005247692.2 | XP_005247749.1 | 8 - 9 | 895 | 62 | ||
| XM_006713721.2 | XP_006713784.1 | 8 - 9 | 905 | 62 | ||
| GenBank mRNA | AF300648.1 | - | 7 - 8 | 694 | 62 | |
| AK001890.1 | - | 8 - 9 | 974 | 62 | ||
| AK290571.1 | - | 8 - 9 | 905 | 62 | ||
| AK297174.1 | - | 8 - 9 | 771 | 62 | ||
| BC000873.1 | - | 8 - 9 | 784 | 3460570 | 62 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03002038.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000465153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000465153.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | ENST00000595265.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036082.hg.1 | TC03002038.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036082.hg.1 | ENST00000232564.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036082.hg.1 | ENST00000468623.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03036082.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000465153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000465153.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000466899.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000497513.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | ENST00000595265.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013264.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198832.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198832.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198832.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198832.hg.1 | ENST00000232564.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198832.hg.1 | ENST00000468623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300198832.hg.1 | NM_021629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300198832.hg.1 | TC0300013264.hg.1 | Clariom D TC |

GNB4
Genomic Map