Assay Details
| Gene Symbol: KALRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322989.1 | NP_001309918.1 | 16 - 17 | 2982 | 71 | |
| NM_001322991.1 | NP_001309920.1 | 16 - 17 | 2943 | 71 | ||
| XM_006713811.3 | XP_006713874.1 | 16 - 17 | 3227 | 71 | ||
| XM_006713813.3 | XP_006713876.1 | 16 - 17 | 3188 | 71 | ||
| XM_011513281.2 | XP_011511583.1 | 16 - 17 | 25876 | 71 | ||
| XM_017007433.1 | XP_016862922.1 | 16 - 17 | 3230 | 71 | ||
| XM_017007435.1 | XP_016862924.1 | 16 - 17 | 3191 | 71 | ||
| GenBank mRNA | AK314126.1 | - | 16 - 17 | 2982 | 71 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

KALRN
Genomic Map