Assay Details
| Gene Symbol: KALRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024660.4 | NP_001019831.2 | 52 - 53 | 7566 | 113 | |
| NM_001322993.1 | NP_001309922.1 | 19 - 20 | 2478 | 113 | ||
| NM_007064.4 | NP_008995.2 | 19 - 20 | 2481 | 113 | ||
| XM_006713810.3 | XP_006713873.1 | 52 - 53 | 7811 | 113 | ||
| XM_006713811.3 | XP_006713874.1 | 52 - 53 | 7784 | 113 | ||
| XM_006713812.3 | XP_006713875.1 | 52 - 53 | 7772 | 113 | ||
| XM_006713813.3 | XP_006713876.1 | 52 - 53 | 7745 | 113 | ||
| XM_006713814.3 | XP_006713877.1 | 51 - 52 | 7715 | 113 | ||
| XM_011513279.2 | XP_011511581.1 | 52 - 53 | 7808 | 113 | ||
| XM_011513280.2 | XP_011511582.1 | 52 - 53 | 7809 | 113 | ||
| XM_011513281.2 | XP_011511583.1 | 52 - 53 | 30433 | 113 | ||
| XM_017007430.1 | XP_016862919.1 | 51 - 52 | 7712 | 113 | ||
| GenBank mRNA | AB011422.1 | - | 19 - 20 | 2435 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

KALRN
Genomic Map