Assay Details
| Gene Symbol: LAMC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002293.3 | NP_002284.3 | 4 - 5 | 1280 | 74 | |
| GenBank mRNA | CN388644.1 | - | 2 - 3 | 192 | 74 | |
| J03202.1 | - | 4 - 5 | 1282 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001583.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein complex assembly
cell adhesion
endoderm development
cell migration
extracellular matrix disassembly
extracellular matrix organization
hemidesmosome assembly
substrate adhesion-dependent cell spreading
positive regulation of epithelial cell proliferation
extracellular matrix structural constituent
protein complex assembly
cell adhesion
endoderm development
cell migration
extracellular matrix disassembly
extracellular matrix organization
hemidesmosome assembly
substrate adhesion-dependent cell spreading
positive regulation of epithelial cell proliferation
extracellular matrix structural constituent
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01024490.hg.1 | TC01001583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024490.hg.1 | ENST00000258341.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024490.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024492.hg.1 | TC01001583.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024492.hg.1 | ENST00000258341.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01024492.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179845.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179845.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179845.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179845.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179845.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179845.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179845.hg.1 | TC0100010863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100179843.hg.1 | TC0100010863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000258341.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000466964.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000478064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000479499.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000484114.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | ENST00000495918.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100010863.hg.1 | NM_002293.3 | Refseq |

LAMC1
Genomic Map