Assay Details
Gene Symbol: LINC01606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CA432459.1 | - | 1 - 2 | 219 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000507074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000507074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000507074.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000507074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000507074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000507074.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000517611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000518556.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519160.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519241.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000519314.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000520929.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000521653.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800007715.hg.1 | ENST00000523341.1 | Ensembl |
