Assay Details
| Gene Symbol: LOC101929570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110258.2 | 1 - 2 | 52 | 104 | ||
| GenBank mRNA | BC042876.1 | - | 1 - 2 | 52 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101929570| Gene Symbol: LOC101929570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110258.2 | 1 - 2 | 52 | 104 | ||
| GenBank mRNA | BC042876.1 | - | 1 - 2 | 52 | 104 | |
Set Membership: |
|