Assay Details
| Gene Symbol: CENPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016343.3 | NP_057427.3 | 16 - 17 | 8489 | 151 | |
| XM_011509082.2 | XP_011507384.1 | 15 - 16 | 8320 | 151 | ||
| XM_017000086.1 | XP_016855575.1 | 16 - 17 | 8476 | 151 | ||
| GenBank mRNA | U19769.1 | - | 8773 | 151 | ||
| U25725.1 | - | 4 - 5 | 2026 | 151 | ||
| U30872.1 | - | 16 - 17 | 8384 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001791.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cell cycle
kidney development
chromosome segregation
sister chromatid cohesion
mitotic nuclear division
mitotic spindle assembly checkpoint
muscle organ development
cell proliferation
regulation of G2/M transition of mitotic cell cycle
protein transport
regulation of striated muscle tissue development
ventricular system development
cell differentiation
response to drug
negative regulation of transcription, DNA-templated
cell division
metaphase plate congression
kinetochore assembly
regulation of cell cycle
DNA biosynthetic process
chromatin binding
protein binding
protein C-terminus binding
transcription factor binding
protein homodimerization activity
dynein binding
mitotic cell cycle
kidney development
chromosome segregation
sister chromatid cohesion
mitotic nuclear division
mitotic spindle assembly checkpoint
muscle organ development
cell proliferation
regulation of G2/M transition of mitotic cell cycle
protein transport
regulation of striated muscle tissue development
ventricular system development
cell differentiation
response to drug
negative regulation of transcription, DNA-templated
cell division
metaphase plate congression
kinetochore assembly
regulation of cell cycle
DNA biosynthetic process
chromatin binding
protein binding
protein C-terminus binding
transcription factor binding
protein homodimerization activity
dynein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000467765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000467765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000467765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000467765.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000469862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000469862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000469862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000495259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000495259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000495259.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000614578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000614578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | ENST00000614578.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028020.hg.1 | TC01001791.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028020.hg.1 | ENST00000366955.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028020.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185292.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185292.hg.1 | ENST00000469862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185292.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185292.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185292.hg.1 | ENST00000469862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185292.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185292.hg.1 | TC0100011581.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000366955.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000464322.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000467765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000467765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000467765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000467765.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000469862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000469862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000469862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000495259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000495259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000495259.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000614578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000614578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | ENST00000614578.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011581.hg.1 | NM_016343.3 | Refseq |

CENPF
Genomic Map