Assay Details
| Gene Symbol: KCNMA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014797.2 | NP_001014797.1 | 19 - 20 | 2283 | 120 | |
| NM_001161353.1 | NP_001154825.1 | 18 - 19 | 2271 | 120 | ||
| NM_001271518.1 | NP_001258447.1 | 18 - 19 | 2121 | 120 | ||
| NM_001322832.1 | NP_001309761.1 | 18 - 19 | 2271 | 120 | ||
| NM_001322835.1 | NP_001309764.1 | 19 - 20 | 2283 | 120 | ||
| NM_001322837.1 | NP_001309766.1 | 19 - 20 | 2283 | 120 | ||
| NM_002247.3 | NP_002238.2 | 18 - 19 | 2271 | 120 | ||
| XM_005269776.3 | XP_005269833.1 | 18 - 19 | 2260 | 120 | ||
| XM_005269787.3 | XP_005269844.1 | 18 - 19 | 2260 | 120 | ||
| XM_005269789.2 | XP_005269846.1 | 19 - 20 | 2204 | 120 | ||
| XM_005269796.2 | XP_005269853.1 | 18 - 19 | 2192 | 120 | ||
| XM_006717826.2 | XP_006717889.1 | 18 - 19 | 2192 | 120 | ||
| XM_011539781.2 | XP_011538083.1 | 20 - 21 | 2404 | 120 | ||
| XM_011539782.2 | XP_011538084.1 | 19 - 20 | 2344 | 120 | ||
| XM_017016212.1 | XP_016871701.1 | 20 - 21 | 2404 | 120 | ||
| XM_017016214.1 | XP_016871703.1 | 19 - 20 | 2392 | 120 | ||
| XM_017016216.1 | XP_016871705.1 | 20 - 21 | 2404 | 120 | ||
| XM_017016217.1 | XP_016871706.1 | 19 - 20 | 2344 | 120 | ||
| XM_017016218.1 | XP_016871707.1 | 18 - 19 | 1729 | 120 | ||
| XM_017016222.1 | XP_016871711.1 | 18 - 19 | 2260 | 120 | ||
| GenBank mRNA | AB113575.1 | - | 18 - 19 | 2022 | 120 | |
| AB209488.1 | - | 18 - 19 | 1689 | 121 | ||
| AB524033.1 | - | 18 - 19 | 1944 | 120 | ||
| AF025999.1 | - | 18 - 19 | 2022 | 120 | ||
| AF118141.1 | - | 18 - 19 | 1899 | 120 | ||
| AY040849.1 | - | 18 - 19 | 2036 | 120 | ||
| BC137115.1 | - | 18 - 19 | 2174 | 9021112 | 120 | |
| BC137137.1 | - | 18 - 19 | 2174 | 9021134 | 120 | |
| U11058.2 | - | 18 - 19 | 1985 | 120 | ||
| U11717.1 | - | 18 - 19 | 2046 | 120 | ||
| U13913.1 | - | 18 - 19 | 2172 | 120 | ||
| U23767.1 | - | 18 - 19 | 2094 | 120 | ||
More Information
Set Membership: |
|

KCNMA1
Genomic Map