Assay Details
| Gene Symbol: SKINT1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026749.2 | 5 - 6 | 823 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TraB domain containing 2B | TRABD2B | TIKI2 | -164.4kb | Sense | Chr. 1 - 47760528 - 47997367 on NCBI Build GRCh38 |
| Downstream | solute carrier family 5 member 9 | SLC5A9 | SGLT4 | 57.4kb | Anti-sense | Chr. 1 - 48221113 - 48248644 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002636.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01041209.hg.1 | TC01002636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041209.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041208.hg.1 | TC01002636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041208.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041208.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041214.hg.1 | TC01002636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041214.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041214.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041212.hg.1 | TC01002636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041212.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041212.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000604015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | ENST00000604015.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041210.hg.1 | TC01002636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041210.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041211.hg.1 | TC01002636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041211.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041211.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041213.hg.1 | TC01002636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041213.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041213.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041215.hg.1 | TC01002636.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041215.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01041215.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205632.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205632.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205632.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205632.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205632.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205632.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205632.hg.1 | TC0100014117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100205634.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205634.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205634.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205634.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205634.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205634.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205634.hg.1 | TC0100014117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000604015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | ENST00000604015.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014117.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205635.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205635.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205635.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205635.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205635.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205635.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205635.hg.1 | TC0100014117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100205637.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205637.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205637.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205637.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205637.hg.1 | TC0100014117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100205628.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205628.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205628.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205628.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205628.hg.1 | ENST00000452503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205628.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205628.hg.1 | TC0100014117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100205631.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205631.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205631.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205631.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205631.hg.1 | TC0100014117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100205638.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205638.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205638.hg.1 | ENST00000453388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205638.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205638.hg.1 | TC0100014117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100205629.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205629.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205629.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205629.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205629.hg.1 | TC0100014117.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100205630.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205630.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205630.hg.1 | ENST00000401057.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100205630.hg.1 | NR_026749.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100205630.hg.1 | TC0100014117.hg.1 | Clariom D TC |

SKINT1L
Genomic Map