Assay Details
Gene Symbol: ZNF192P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_103448.1 | 3 - 4 | 399 | 77 | ||
GenBank mRNA | DN994493.1 | - | 2 - 3 | 521 | 77 | |
HY011556.1 | - | 3 - 4 | 399 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600150120.hg.1 | TC0600007403.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | ENST00000570126.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007403.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600150118.hg.1 | TC0600007403.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | ENST00000440790.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | ENST00000562227.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | ENST00000565888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | ENST00000570042.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | NR_103448.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600150119.hg.1 | TC0600007403.hg.1 | Clariom D TC |
