Assay Details
Gene Symbol: DNAJC27-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_034113.1 | 1 - 2 | 120 | 103 | ||
GenBank mRNA | CR933670.1 | - | 1 - 2 | 105 | 103 | |
DA314471.1 | - | 1 - 2 | 120 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000140.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02001616.hg.1 | TC02000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02001616.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001616.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421842.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02001620.hg.1 | TC02000140.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02001620.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02001620.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000421842.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000422449.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000421842.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000422449.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147577.hg.1 | TC0200006982.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421842.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000421904.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000422449.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000428614.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200006982.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000434897.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000445389.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | ENST00000451291.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | NR_034113.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200147583.hg.1 | TC0200006982.hg.1 | Clariom D TC |
