Assay Details
| Gene Symbol: DUSP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319658.1 | NP_001306587.1 | 1 - 2 | 370 | 75 | |
| NM_001319659.1 | NP_001306588.1 | 2 - 3 | 329 | 75 | ||
| NM_017823.4 | NP_060293.2 | 2 - 3 | 322 | 75 | ||
| GenBank mRNA | AB164404.1 | - | 1 - 2 | 355 | 75 | |
| AF086553.1 | - | 1 - 2 | 170 | 75 | ||
| AK000449.1 | - | 2 - 3 | 306 | 75 | ||
| BC001140.1 | - | 2 - 3 | 292 | 3535406 | 75 | |
| BG823348.1 | - | 1 - 2 | 352 | 75 | ||
| BQ679163.1 | - | 2 - 3 | 341 | 75 | ||
| DA428735.1 | - | 1 - 2 | 373 | 75 | ||
More Information
Set Membership: |
|

DUSP23
Genomic Map