Assay Details
| Gene Symbol: RET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020630.4 | NP_065681.1 | 523 | 92 | ||
| NM_020975.4 | NP_066124.1 | 523 | 92 | |||
| GenBank mRNA | AK291807.1 | - | 393 | 92 | ||
| BC003072.2 | - | 513 | 92 | |||
| BC004257.1 | - | 513 | 3160389 | 92 | ||
| X15262.1 | - | 482 | 92 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10000268.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
ureteric bud development
neural crest cell migration
embryonic epithelial tube formation
protein phosphorylation
homophilic cell adhesion via plasma membrane adhesion molecules
neuron cell-cell adhesion
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
posterior midgut development
positive regulation of neuron projection development
positive regulation of neuron maturation
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
positive regulation of cell migration
membrane protein proteolysis
positive regulation of cell adhesion mediated by integrin
ureter maturation
response to drug
neuron maturation
positive regulation of GTPase activity
positive regulation of cell size
positive regulation of transcription, DNA-templated
response to pain
enteric nervous system development
regulation of axonogenesis
retina development in camera-type eye
innervation
Peyer's patch morphogenesis
cellular response to retinoic acid
positive regulation of metanephric glomerulus development
lymphocyte migration into lymphoid organs
positive regulation of serine phosphorylation of STAT3 protein
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor activity
Ras guanyl-nucleotide exchange factor activity
calcium ion binding
protein binding
ATP binding
MAPK cascade
ureteric bud development
neural crest cell migration
embryonic epithelial tube formation
protein phosphorylation
homophilic cell adhesion via plasma membrane adhesion molecules
neuron cell-cell adhesion
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
posterior midgut development
positive regulation of neuron projection development
positive regulation of neuron maturation
peptidyl-tyrosine phosphorylation
regulation of cell adhesion
positive regulation of cell migration
membrane protein proteolysis
positive regulation of cell adhesion mediated by integrin
ureter maturation
response to drug
neuron maturation
positive regulation of GTPase activity
positive regulation of cell size
positive regulation of transcription, DNA-templated
response to pain
enteric nervous system development
regulation of axonogenesis
retina development in camera-type eye
innervation
Peyer's patch morphogenesis
cellular response to retinoic acid
positive regulation of metanephric glomerulus development
lymphocyte migration into lymphoid organs
positive regulation of serine phosphorylation of STAT3 protein
positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor activity
Ras guanyl-nucleotide exchange factor activity
calcium ion binding
protein binding
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000340058.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000355710.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000479913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000479913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000479913.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000498820.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000498820.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000498820.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000498820.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000498820.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000498820.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | ENST00000615310.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020630.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000007403.hg.1 | NM_020975.5 | Refseq |

RET
Genomic Map