Assay Details
| Gene Symbol: BOD1L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257964.1 | NP_001244893.1 | 1 | 2049 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336875 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336831 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735060 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735079 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735008 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072493 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735045 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735085 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735041 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735075 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735047 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336843 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336847 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735059 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336792 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735073 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735009 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336932 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734994 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735027 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336806 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734966 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072495 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336931 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336876 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735083 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734982 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072497 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072491 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734968 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072473 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072490 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072479 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735014 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336856 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072470 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336871 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735022 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072486 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336904 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336878 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336895 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734997 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735046 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735026 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336908 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734959 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734961 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734969 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735077 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336793 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735019 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735051 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734987 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336897 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734972 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336845 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734993 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336893 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735076 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734964 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734978 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336905 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735002 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072489 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072500 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735044 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734975 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336858 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735069 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734990 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735042 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735066 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336886 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336798 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336870 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735057 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336834 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735087 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735089 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734998 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336917 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734974 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072498 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336820 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735082 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336827 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735025 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336814 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336817 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072481 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735081 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17072472 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735064 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336844 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735024 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336935 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735030 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10734970 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735010 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336918 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10735054 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336836 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P31336818 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3503284 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P345212 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 34493 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3054 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3503284 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3503284 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10735087 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10735089 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10734959 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31336935 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10735086 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10734958 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31336935 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10735087 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10735086 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10735089 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10734958 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10734959 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P345212 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P345212 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8021347 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8021345 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8021348 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8021346 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8021344 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 231368_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11763338_PM_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11763338_a_at | - | - |
| Affymetrix Human Clariom D Assay | PSR1800150697.hg.1 | NM_001257964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1800150697.hg.1 | TC1800007392.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1800150697.hg.1 | NM_001257964.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 231368_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 231368_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 231368_at6 | - | - |
| Human Genome U95 Set | 47609_at | - | - |
| Human Genome U95 Set | 47609_PM_at | - | - |
| Human Genome U95 Set | 74189_PM_at | - | - |
| Human Genome U95 Set | 74189_at | - | - |
| Human Genome U133 Set | 231368_at | - | - |
| Human Genome U133 Set | 231368_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010674 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A010674 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 222998 | - | - |

BOD1L2
Genomic Map