Assay Details
Gene Symbol: C1orf100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012970.2 | NP_001012988.1 | 3 - 4 | 268 | 110 | |
XM_011544124.2 | XP_011542426.1 | 4 - 5 | 1110 | 110 | ||
GenBank mRNA | HY055397.1 | - | 3 - 4 | 224 | 110 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001987.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01030771.hg.1 | TC01001987.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01030771.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030771.hg.1 | ENST00000366537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030771.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030771.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030769.hg.1 | TC01001987.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01030769.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030769.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030769.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030768.hg.1 | TC01001987.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01030768.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030768.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100012219.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01030770.hg.1 | TC01001987.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01030770.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030770.hg.1 | ENST00000366537.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030770.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01030770.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189934.hg.1 | TC0100012219.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012219.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189933.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189933.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189933.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189933.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189933.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189933.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189933.hg.1 | TC0100012219.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | ENST00000366537.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | ENST00000470211.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | ENST00000486803.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | NM_001276348.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | NM_001276349.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189935.hg.1 | TC0100012219.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100189932.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189932.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189932.hg.1 | ENST00000308105.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100189932.hg.1 | NM_001012970.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100189932.hg.1 | TC0100012219.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 191591 | - | - |
