Assay Details
| Gene Symbol: SPON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006108.3 | NP_006099.2 | 14 - 15 | 2266 | 60 | |
| GenBank mRNA | AB018305.2 | - | 13 - 14 | 1751 | 60 | |
| AB051390.1 | - | 14 - 15 | 2022 | 60 | ||
| AK001584.1 | - | 6 - 7 | 835 | 60 | ||
| AK074803.1 | - | 14 - 15 | 2212 | 60 | ||
| AK291780.1 | - | 14 - 15 | 2214 | 60 | ||
| BC019825.1 | - | 3 - 4 | 356 | 60 | ||
| BC041974.1 | - | 2 - 3 | 349 | 60 | ||
| BC136513.1 | - | 14 - 15 | 2208 | 9020503 | 60 | |
| BC136563.1 | - | 14 - 15 | 2208 | 9020553 | 60 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000211.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002850.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002850.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002850.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11002850.hg.1 | TC11000211.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100148670.hg.1 | TC1100006911.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000310358.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000526298.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000533633.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000576479.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | ENST00000591785.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006911.hg.1 | NM_006108.3 | Refseq |

SPON1
Genomic Map