Assay Details
| Gene Symbol: IQCA1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001304419.1 | NP_001291348.1 | 1 - 2 | 48 | 111 | |
| GenBank mRNA | AL045644.2 | - | 1 - 2 | 48 | 111 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001304419.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194244.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194244.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194244.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194244.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194244.hg.1 | TC0700013064.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194245.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194245.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194245.hg.1 | ENST00000615129.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700194245.hg.1 | NM_001304419.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700194245.hg.1 | TC0700013064.hg.1 | Clariom D TC |

IQCA1L
Genomic Map