Assay Details
| Gene Symbol: AP5M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026895.1 | 1 - 2 | 499 | 88 | ||
| GenBank mRNA | DB204683.1 | - | 1 - 2 | 361 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | exocyst complex component 5 | EXOC5 |
HSEC10, PRO1912, SEC10, SEC10L1, SEC10P |
-166.0 | Anti-sense | Chr. 14 - 57202476 - 57268899 on NCBI Build GRCh38 |
| Downstream |
N(alpha)-acetyltransferase 30, NatC catalytic subunit |
NAA30 |
C14orf35, MAK3, Mak3p, NAT12, NAT12P |
110.1kb | Sense | Chr. 14 - 57390553 - 57412748 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000340.hg.1
Set Membership: |
|

AP5M1
Genomic Map