Assay Details
| Gene Symbol: BIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320641.1 | NP_001307570.1 | 12 - 13 | 1534 | 86 | |
| NM_001320642.1 | NP_001307571.1 | 13 - 14 | 2121 | 86 | ||
| NM_139343.2 | NP_647593.1 | 13 - 14 | 1627 | 86 | ||
| NM_139344.2 | NP_647594.1 | 12 - 13 | 1498 | 86 | ||
| XM_005263642.3 | XP_005263699.1 | 14 - 15 | 1745 | 86 | ||
| XM_005263643.3 | XP_005263700.1 | 13 - 14 | 1652 | 86 | ||
| XM_005263645.2 | XP_005263702.1 | 13 - 14 | 1136 | 86 | ||
| XM_006712427.2 | XP_006712490.1 | 12 - 13 | 1091 | 86 | ||
| XM_011510975.2 | XP_011509277.1 | 14 - 15 | 2378 | 86 | ||
| XM_017003819.1 | XP_016859308.1 | 14 - 15 | 1559 | 86 | ||
| GenBank mRNA | AF004015.1 | - | 13 - 14 | 1243 | 86 | |
| AF043901.1 | - | 12 - 13 | 1114 | 86 | ||
| AF070576.1 | - | 9 - 10 | 849 | 86 | ||
| AL713697.1 | - | 13 - 14 | 1588 | 86 | ||
| BE047931.1 | - | 4 - 5 | 432 | 86 | ||
| U87558.1 | - | 13 - 14 | 1264 | 86 | ||
More Information
Set Membership: |
|

BIN1
Genomic Map