Assay Details
Gene Symbol: KCNC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001112741.1 | NP_001106212.1 | 3 - 4 | 1738 | 97 | |
GenBank mRNA | AK123157.1 | - | 1 - 2 | 463 | 97 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11000233.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
potassium ion transport
response to toxic substance
response to light intensity
response to auditory stimulus
response to amine
cerebellum development
globus pallidus development
cellular response to drug
response to potassium ion
protein homooligomerization
protein tetramerization
response to fibroblast growth factor
potassium ion transmembrane transport
positive regulation of potassium ion transmembrane transport
response to nerve growth factor
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
ion channel binding
potassium ion transport
response to toxic substance
response to light intensity
response to auditory stimulus
response to amine
cerebellum development
globus pallidus development
cellular response to drug
response to potassium ion
protein homooligomerization
protein tetramerization
response to fibroblast growth factor
potassium ion transmembrane transport
positive regulation of potassium ion transmembrane transport
response to nerve growth factor
voltage-gated potassium channel activity
delayed rectifier potassium channel activity
ion channel binding
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11003102.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003102.hg.1 | TC11000233.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11003101.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11003101.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11003101.hg.1 | TC11000233.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000525802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000525802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000525802.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149159.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149159.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149159.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149159.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149159.hg.1 | TC1100006977.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000379472.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000525802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000525802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000525802.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1100006977.hg.1 | NM_004976.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149157.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149157.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149157.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149157.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149157.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149157.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149157.hg.1 | TC1100006977.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100149158.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149158.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149158.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149158.hg.1 | ENST00000265969.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149158.hg.1 | ENST00000526029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100149158.hg.1 | NM_001112741.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100149158.hg.1 | TC1100006977.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 172114 | - | - |
