| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002729.hg.1 |
TC05000220.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002729.hg.1 |
ENST00000256759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002729.hg.1 |
ENST00000396947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002729.hg.1 |
ENST00000511025.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002729.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002729.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002728.hg.1 |
TC05000220.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002728.hg.1 |
ENST00000256759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002728.hg.1 |
ENST00000396947.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002728.hg.1 |
ENST00000511025.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002728.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05002728.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149868.hg.1 |
TC0500007376.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500149869.hg.1 |
TC0500007376.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000256759.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000396947.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000491717.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000497789.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
ENST00000504226.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_006350.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500007376.hg.1 |
NM_013409.2 |
Refseq |