Assay Details
| Gene Symbol: FST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006350.3 | NP_006341.1 | 3 - 4 | 856 | 61 | |
| NM_013409.2 | NP_037541.1 | 3 - 4 | 856 | 61 | ||
| XM_005248400.4 | XP_005248457.1 | 3 - 4 | 951 | 61 | ||
| XM_005248401.4 | XP_005248458.1 | 3 - 4 | 949 | 61 | ||
| XM_005248402.4 | XP_005248459.1 | 3 - 4 | 952 | 61 | ||
| XM_005248403.4 | XP_005248460.1 | 3 - 4 | 950 | 61 | ||
| XM_011543099.2 | XP_011541401.1 | 3 - 4 | 666 | 61 | ||
| XM_017008954.1 | XP_016864443.1 | 3 - 4 | 952 | 61 | ||
| XM_017008955.1 | XP_016864444.1 | 3 - 4 | 1044 | 61 | ||
| GenBank mRNA | AB451330.1 | - | 3 - 4 | 498 | 61 | |
| AB451474.1 | - | 3 - 4 | 498 | 61 | ||
| BC004107.2 | - | 3 - 4 | 703 | 3688745 | 61 | |
| CR541813.1 | - | 3 - 4 | 498 | 61 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05000220.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
hematopoietic progenitor cell differentiation
signal transduction
negative regulation of activin receptor signaling pathway
positive regulation of hair follicle development
signal transducer activity
protein binding
heparan sulfate proteoglycan binding
activin binding
negative regulation of transcription from RNA polymerase II promoter
hematopoietic progenitor cell differentiation
signal transduction
negative regulation of activin receptor signaling pathway
positive regulation of hair follicle development
signal transducer activity
protein binding
heparan sulfate proteoglycan binding
activin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR05002730.hg.1 | TC05000220.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002730.hg.1 | ENST00000256759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002730.hg.1 | ENST00000396947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002730.hg.1 | ENST00000511025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002730.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002730.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002731.hg.1 | TC05000220.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002731.hg.1 | ENST00000256759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002731.hg.1 | ENST00000396947.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002731.hg.1 | ENST00000511025.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002731.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05002731.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000497789.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500007376.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | ENST00000491717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149870.hg.1 | TC0500007376.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | ENST00000256759.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | ENST00000396947.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | ENST00000504226.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | NM_006350.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | NM_013409.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500149871.hg.1 | TC0500007376.hg.1 | Clariom D TC |

FST
Genomic Map