Assay Details
| Gene Symbol: MALT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006785.3 | NP_006776.1 | 9 - 10 | 1272 | 182 | |
| NM_173844.2 | NP_776216.1 | 8 - 9 | 1239 | 149 | ||
| XM_011525794.1 | XP_011524096.1 | 9 - 10 | 1238 | 182 | ||
| GenBank mRNA | AB026118.1 | - | 8 - 9 | 1046 | 149 | |
| AF130356.2 | - | 9 - 10 | 1178 | 182 | ||
| AF316597.1 | - | 9 - 10 | 1272 | 182 | ||
| AK291386.1 | - | 9 - 10 | 1238 | 182 | ||
| BC030143.2 | - | 8 - 9 | 1093 | 4765195 | 149 | |
More Information
Set Membership: |
|

MALT1
Genomic Map